Learn More
Thermo Scientific™ T4 DNA Polymerase Promotion Available on GSA/VA Contract for Federal Government customers only.
Ensure accuracy in DNA synthesis with template-dependent T4 DNA Polymerase that catalyzes the 5'-3' synthesis from primed single-stranded DNA.
Manufacturer:  Thermo Scientific™ EP0062
 View more versions of this product
Catalog No. FEREP0062PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
T4 DNA Polymerase, a template-dependent DNA polymerase, catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.
Highlights
  • Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I, E. coli,and Klenow fragment
  • Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase buffers
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Blunting of DNA ends: fill-in 5'-overhangs or/and removal of 3'-overhangs (1, 2); Blunting of PCR products with 3'-dA overhangs; Synthesis of label
2 x 1mL of 5X Reaction Buffer
T4 DNA Polymerase
500U