Learn More
Simport™ Scientific T500 Screw Caps for Internally Threaded Sample Tubes
Simport™ Scientific T500 Screw Caps for Internally Threaded Sample Tubes available with silicone washer seal.
$236.50
Specifications
Product Type Screw Cap
For Use With T500 series Sample tube
Autoclavable Yes
Pre-sterilized No
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
23-045-265 View Documents Simport™ Scientific
T500NOS
Natural Pack of 1000 for $236.50
Add to cart
23-045-266 View Documents Simport™ Scientific
T500BOS
Blue Pack of 1000 for $236.50
Add to cart
Description
Description
  • Made of polypropylene
  • With silicone washer seal
  • Autoclavable
  • For internally threaded tubes
  • Tubes sold separately
Specifications
Specifications
Screw Cap
Yes
Polypropylene
1000
T500 series Sample tube
No
Silicone