Tocris™ TA 01

Induces cardiomyocyte differentiation from hESCs

Manufacturer: Tocris™ 5383/50

 View more versions of this product

Catalog No. 53-835-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors TA 01
    Purity >98
    Molecular Weight 351.32
    Quantity 50mg
    Product Type TA 01

    Induces cardiomyocyte differentiation from human embryonic stem cells following mesoderm induction at 1µM. Inhibits cardiomyocyte differentiation at 5µM. Potently inhibits CK1ε, p38α, and CK1δ (IC50 values are 6.4, 6.7 and 6.8nM respectively).

    • Chemical Name: 4-(2-(2,6-Difluorophenyl)-4-(fluorophenyl)-1H-imidazol-5-yl)pyridine
    • Formula: C20H12F3N3
    • Solubility: Soluble to 100mM in DMSO and to 10mM in ethanol with gentle warming
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit