Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ TA 02

Induces cardiomyocyte differentiation from hESCs

Manufacturer:  Tocris™ 5384/10

 View more versions of this product

Catalog No. 53-841-0


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Induces cardiomyocyte differentiation from human embryonic stem cells following mesoderm induction. Potently inhibits p38α, CK1δ and CK1ε (IC50 values are 20, 32 and 32nM respectively).
  • Chemical Name: 4-(2-(2-Fluorophenyl)-4-(fluorophenyl)-1H-imidazol-5-yl)pyridine
  • Formula: C20H13F2N3
  • Solubility: Soluble to 100mM in DMSO and to 20mM in ethanol
  • Purity: >98
Description & Specifications

Specifications

TA 02
>98
10mg
TA 02
333.33
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit