Learn More
TAAR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00114W
Catalog No. PIOSR00114W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material.
Orphan receptor. Could be a receptor for trace amines. Of the biogenic trace amine tested, activated only by beta-phenylethylamine (beta-PEA) and tryptamine; however, the response is of low potency. Trace amines are biogenic amines present in very low levels in mammalian tissues. Although some trace amines have clearly defined roles as neurotransmitters in invertebrates, the extent to which they function as true neurotransmitters in vertebrates has remained speculative. Trace amines are likely to be involved in a variety of physiological functions that have yet to be fully understood. This receptor seems to be mediated by the G(s)-class of G-proteins which activate adenylate cyclase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAAR4
Polyclonal
Taar4
Ta2, Gm226, TAAR 4
Rabbit
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q5QD15, Q923Y7
TAAR4
A synthetic peptide from c-terminal region of rat trace amine-associated receptor 4
RUO
209513, 294122
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Recommendations
Recommendations