Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Table Clamp

For securing and holding a rod up to 12.5mm dia. vertically in a machined V-groove.

Manufacturer:  Eisco™ CH0688F/DEL

Catalog No. 04-999-131

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

Clamps to tabletop up to 65mm thick.
Specifications

Specifications

Table Clamp
5.9 in.
14.98 cm
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit