Learn More
American Educational Products Pulley Cord for Table Clamp
A critical part of a physic table pulley demonstration.
$20.32
Specifications
For Use With (Application) | Easy height adjustment for table-edge demos |
---|
Catalog Number | Mfr. No. | Item Description | Weight (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Item Description | Weight (English) | Price | Quantity & Availability | ||||
S40883
![]() |
American Educational Products
1TB80 |
137m (450 ft.) length pulley | 80 lb. |
Each for $20.32
|
|
Description
A gripping twosome for easily adjusting the height of a pulley for table-edge demos.
- 5cm diameter plastic pulley, mounted on a 23.5cm long metal shaft with a hardened-steel table clamp
- Pulley shaft is held in place with a clamping screw
Order Info
Optional accessories include a 1mm-thick, 80-lb. test and 10-lb. test pulley cord made of quality braided nylon fishing line
Specifications
Easy height adjustment for table-edge demos |