Learn More
TACC2 Mouse anti-Human, Clone: OTI4D10, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF804146
Catalog No. 50-166-6831
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Transforming acidic coiled-coil proteins are a conserved family of centrosome- and microtubule-interacting proteins that are implicated in cancer. This gene encodes a protein that concentrates at centrosomes throughout the cell cycle. This gene lies within a chromosomal region associated with tumorigenesis. Expression of this gene is induced by erythropoietin and is thought to affect the progression of breast tumors. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
TACC2
Monoclonal
Unconjugated
TACC2
Lyophilized
TACC2
Human recombit protein fragment corresponding to amino acids 727-1026 of human TACC2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI4D10
PBS with 8% trehalose and no preservative; pH 7.3
O95359
AZU-1, ECTACC
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10579