Learn More
TACO1/CCDC44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 211471AP
Catalog No. 50-173-3280
Add to cart
Description
Description
TACO1 is a mitochondrial translational activator required for efficient translation of cytochrome c oxidase (Weraarpachai et al., 2009), a component of complex IV of the mitochondrial respiratory complex.
Specifications
Specifications
TACO1/CCDC44
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
B2RYT9, Q8K0Z7, Q9BSH4
CCDC44, TACO1
Rabbit
IgG
150 μL
-20°C
Primary
360645, 51204, 70207
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TACO1
Liquid
Taco1
TACO1/CCDC44 Fusion Protein Ag15688
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat