Learn More
Selleck Chemical LLCSupplier Diversity Partner Tadalafil 500mg 171596-29-5 IC351  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S1512500MG
Tadalafil (IC351) is a PDE-5 inhibitor with IC50 of 1.8 nM in a cell-free assay. Tadalafil is at least 9000 times more selective for PDE5 than most of the other families of PDEs, with the exception of PDE11. It can partial inhibits PDE11
Catalog No. 50-635-6