Learn More
TAF13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 246721AP
Catalog No. 50-173-4483
Add to cart
Description
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a small subunit associated with a subset of TFIID complexes. This subunit interacts with TBP and with two other small subunits of TFIID, TAF10 and TAF11. There is a pseudogene located on chromosome 6.
Specifications
Specifications
TAF13
0.6 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P61216, Q15543
TAF(II)18, TAF13, TAF2K, TAFII 18, TAFII18
Rabbit
IgG
150 μL
-20°C
Primary
310784, 6884, 99730
Western Blot
Unconjugated
TAF13
Liquid
Taf13
TAF13 Fusion Protein Ag18193
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat