Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF15 Mouse anti-Human, Mouse, Rat, DyLight™ 550, Clone: 4D71, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192685R

Catalog No. NBP192685R


Add to cart

Description

Description

TAF15 Monoclonal antibody specifically detects TAF15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

TAF15
4D71
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
TAF15
Full length recombinant human TAF15 expressed in and purified from E. coli. [UniProtNo. Q92804]
Protein G purified
DNA Repair, Neuroscience
Primary
8148.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
68kDa, hTAFII68, Npl3, RBP56TAFII68, RNA-binding protein 56, TAF(II)68, TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2NRBP56/CSMF fusion, TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD(RNA-binding protein 56), TATA box-binding protein-associated factor 2N (RNA-binding protein 56), TATA-binding protein-associated factor 2N, TBP-associated factor 15,68 kDa TATA-binding protein-associated factor
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit