Learn More
Aviva Systems Biology Corp TAF1A Antibody - N-terminal region (P100801_T100) | 100uL
Supplier:  Aviva Systems Biology Corp P100801_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TAF1A. Alternate names: SL1, RAFI48, TAFI48, MGC:17061ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62718