Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAF1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549295
Description
Predicted to react with monkey based on sequence homology.
TAF1B Polyclonal antibody specifically detects TAF1B in Human samples. It is validated for Western Blot.Specifications
TAF1B | |
Polyclonal | |
Unconjugated | |
TAF1B | |
4930408G01Rik; A230108M10Rik; MGC:9349; p63; RAF1B; RAFI63; RNA polymerase I-specific TBP-associated factor 63 kDa; RNA polymerase I-specific TBP-associated factor 68 kDa; SL1; SL1, 63kD subunit; Taf1b; TAFI63; TAFI68; Tafi86; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, B; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kDa; TATA box binding protein associated factor 1B; TATA box-binding protein-associated factor 1B; TATA box-binding protein-associated factor RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I, B; TBP-associated factor 1B; TBP-associated factor RNA polymerase I 68; transcription factor TAFI68; transcription initiation factor SL1/TIF-IB subunit B | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
9014 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.7 mg/mL | |
PBS with 0.09% sodium azide | |
Q53T94 | |
TAF1B | |
KLH conjugated synthetic peptide between 367-396 amino acids from the Central region of human TAF1B. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction