Learn More
Aviva Systems Biology Corp TAF3 Antibody - middle region (ARP51249_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51249_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TAF3. Alternate names: MGC25063, TAF140, TAFII140, TAFII-140ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60527