Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF4B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548337

Catalog No. PA548337

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Cell type-specific subunit of the general transcription factor TFIID that may function as a gene-selective coactivator in certain cells. TFIID is a multimeric protein complex that plays a central role in mediating promoter responses to various activators and repressors. TAF4B is a transcriptional coactivator of the p65/RELA NF-kappa-B subunit. Involved in the activation of a subset of antiapoptotic genes including TNFAIP3. May be involved in regulating folliculogenesis. Through interaction with OCBA/POU2AF1, acts as a coactivator of B-cell-specific transcription. Plays a role in spermiogenesis and oogenesis.
Specifications

Specifications

TAF4B
Polyclonal
PBS with 0.09% sodium azide
Q92750
Transcription initiation factor TFIID subunit 4B, Transcription initiation factor TFIID 105 kDa subunit, TAF(II)105, TAFII-105, TAFII105, TAF4B, TAF2C2, TAFII105
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6875
Flow Cytometry, Western Blot
Unconjugated
TAF4B
Liquid
TAF4B
KLH conjugated synthetic peptide between 647-675 amino acids from the C-terminal region of human TAF4B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit