Learn More
TAF5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 192741AP
Catalog No. 50-173-2380
Add to cart
Description
Description
TAF5L (TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L), also known as PAF65B, is a 589 amino acid protein that localizes to the nucleus and contains six WD repeats. Existing as a component of the PCAF histone acetylase complex, TAF5L interacts with a number of TAFs and, via this interaction, plays a role in histone acetylation and transcription initiation. Defects in the gene encoding TAF5L, which is expressed as multiple alternatively spliced isoforms, are associated with the pathogenesis of type I diabetes.
Specifications
Specifications
TAF5L
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75529, Q91WQ5
PAF65 beta, PAF65B, PCAF associated factor 65 beta, TAF5L
Rabbit
IgG
150 μL
-20°C
Primary
102162, 27097, 307927
Western Blot
Unconjugated
TAF5L
Liquid
Taf5l
TAF5L Fusion Protein Ag5942
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat