Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAF7L Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549303

Catalog No. PA549303

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Probably functions as a spermatogensis-specific component of the DNA-binding general transcription factor complex TFIID, a multimeric protein complex that plays a central role in mediating promoter responses to various activators and repressors. May play a role in spermatogenesis.
Specifications

Specifications

TAF7L
Polyclonal
PBS with 0.09% sodium azide
Q5H9L4
Transcription initiation factor TFIID subunit 7-like, Cancer/testis antigen 40, CT40, RNA polymerase II TBP-associated factor subunit Q, TATA box-binding protein-associated factor 50 kDa, Transcription initiation factor TFIID 50 kDa subunit, TAF7L, TAF2Q
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54457
Western Blot
Unconjugated
TAF7L
Liquid
TAF7L
KLH conjugated synthetic peptide between 68-96 amino acids from the N-terminal region of human TAF7L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit