Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAFA3/FAM19A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | TAFA3/FAM19A3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
TAFA3/FAM19A3 Polyclonal specifically detects TAFA3/FAM19A3 in Human samples. It is validated for Western Blot.Specifications
TAFA3/FAM19A3 | |
Polyclonal | |
Rabbit | |
Q2M1P9 | |
284467 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
Chemokine-like protein TAFA-3, family with sequence similarity 19 (chemokine (C-C motif)-like), member A3, MGC138473, TAFA3, TAFA-3 | |
FAM19A3 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title