Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAG-72, anti-Human, Clone: B72.3 (Satumomab), Abnova™

Monoclonal Antibody

Supplier:  Abnova Corporation RAB00001

Catalog No. 89-032-344


Add to Cart

Description

Description

Specifications

Specifications

TAG-72
Monoclonal
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
200 μg
Primary
Human
Antibody
IgG1 κ
Flow Cytometry, Immunohistochemistry
B72.3 (Satumomab)
Flow Cytometry (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
Original antibody is raised against native purified human TAG-72.
RUO
This antibody recognizes human tumor associated glycoprotein 72 (TAG-72).
Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit