Learn More
TAG-72/CA 72-4 Mouse anti-Human, Clone: B72.3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF190272

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Cancer Antigen 72-4 (CA 72-4, TAG 72-4) is a tumor marker that may be used to diagnose and monitor neoplastic diseases.Specifications
TAG-72/CA 72-4 | |
Monoclonal | |
Unconjugated | |
CA72-4 | |
Membrane-enriched fraction of a human breast carcinoma liver metastasis | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin) | |
B72.3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.