Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAG-72 Mouse anti-Human, Rat, Bovine, Canine, Allophycocyanin/Cy7, Clone: CC49, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP233128APCCY7
Description
TAG-72 Monoclonal antibody specifically detects TAG-72 in Human, Rat, Bovine, Canine samples. It is validated for Flow Cytometry.Specifications
TAG-72 | |
CC49 | |
Flow Cytometry | |
Conjugated, Purified | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Primary | |
Recognizes an oncofetal antigen of 220kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. This MAb defines the mucin-carried sialylated-Tn epitope. TAG-72 is usually expressed by adenocarcinomas, but is negative in mesotheliomas. Studies have reported that this antibody has 80% sensitivity and 93% specificity for pulmonary adenocarcinoma. Therefore, TAG-72 is a useful marker to distinguish between mesothelioma and adenocarcinoma. However, false positive reactions can occur so results must be interpreted with the utmost caution. This antibody may be useful in the differentiation of non-small cell carcinomas from small cell carcinomas of the lung. The combined use of anti-TAG-72 and anti-GCDFP-15 is valuable in the diagnosis of apocrine carcinoma. |
Flow Cytometry | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72 | |
Purified TAG-72 protein | |
Protein A purified | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
Bovine, Canine, Human, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title