Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAG-72 Mouse anti-Human, Rat, Bovine, Canine, Hamster, Clone: rB72.3, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen TAG-72
Clone rB72.3
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122843
View Documents
Novus Biologicals
NBP30768510UG
100 μg
Each for $374.34
Add to cart
 
NB122844
View Documents
Novus Biologicals
NBP30768520UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

TAG-72 Monoclonal antibody specifically detects TAG-72 in Human, Rat, Bovine, Canine, Hamster samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TAG-72
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
Membrane-enriched fraction of a human breast carcinoma liver metastasis
Monoclonal
Protein A or G purified
rB72.3
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Human, Rat, Bovine, Canine, Hamster
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit