Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAG-72 Rabbit anti-Human, Alexa Fluor™ 750, Clone: B72.3 (Satumomab), Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252672AF750

Catalog No. NP22672A70


Add to cart

Description

Description

TAG-72 Monoclonal antibody specifically detects TAG-72 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

TAG-72
B72.3 (Satumomab)
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
Rabbit
IgG Kappa
0.1 mL
Primary
Recognises human Tumor associated glycoprotein 72.
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72
Tumour-associated glycoprotein 72 is a glycoprotein found on the surface of many cancer cells, including ovary, breast, colon, and pancreatic cells.
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit