Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAG-72 Rabbit anti-Human, Clone: B72.3 (Satumomab), Novus Biologicals™
Rabbit Monoclonal Antibody
$229.00 - $516.00
Specifications
Antigen | TAG-72 |
---|---|
Clone | B72.3 (Satumomab) |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25267225
|
Novus Biologicals
NBP2526720.025MG |
0.025 mg |
Each for $229.00
|
|
NBP25267202
|
Novus Biologicals
NBP2526720.2MG |
0.2 mg |
Each for $516.00
|
|
Description
TAG-72 Monoclonal specifically detects TAG-72 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.Specifications
TAG-72 | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
182875 | |
IgG | |
Protein A or G purified | |
Recognises human Tumor associated glycoprotein 72. |
B72.3 (Satumomab) | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF | |
Unconjugated | |
Rabbit | |
Human | |
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72 | |
Tumour-associated glycoprotein 72 is a glycoprotein found on the surface of many cancer cells, including ovary, breast, colon, and pancreatic cells. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title