Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAG-72 Rabbit anti-Human, Rat, Bovine, Canine, Hamster, Clone: CA72/2869R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen TAG-72
Clone CA72/2869R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122872
View Documents
Novus Biologicals
NBP30769920UG
20 μg
Each for $159.82
Add to cart
 
NB122871
View Documents
Novus Biologicals
NBP30769910UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

TAG-72 Monoclonal antibody specifically detects TAG-72 in Human, Rat, Bovine, Canine, Hamster samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TAG-72
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
10 mM PBS with 0.05% BSA
Purified TAG-72 protein
Monoclonal
Protein A or G purified
CA72/2869R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Human, Rat, Bovine, Canine, Hamster
CA 72-4, Protein TAG-72, tag 72, tag-72 Protein TAG-72
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit