Learn More
Brady™ Tag Stock - ANSI Caution Header
Identify facility equipment to your exact specifications. Brady™ Tags Stock - ANSI Caution Header are durable one-sided tags that are an economical and convenient way to ID equipment.
Manufacturer:  Brady™ B30255551ANSICA
Catalog No. 19-113-110
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
OSHA 1910.145
Matte
Rectangle
Polyester
Black/yellow on white
100/Roll
-40°F to 193°F