Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAGLN Mouse anti-Human, Clone: SPM606, Abnova™

Mouse monoclonal antibody raised against full length recombinant human TAGLN.

Manufacturer:  Abnova Corporation MAB13163

Catalog No. 89-934-985


Add to cart

Description

Description

The protein encoded by this gene is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Specifications

Specifications

TAGLN
SPM606
Mouse monoclonal antibody raised against full length recombinant human TAGLN.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
DKFZp686P11128/SM22/SMCC/TAGLN1/WS3-10
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
This monoclonal antibody identified as TAGLN, also designated SM22-alpha. It may cross-react with SM22-beta.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
TAGLN
Recombinant protein corresponding to full length human TAGLN.
Protein A/G purification
RUO
Primary
6876
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit