Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAGLN Mouse anti-Human, Clone: TAGLN/247, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14610

Catalog No. 89-034-693


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human TAGLN.

  • High specificity
  • High reactivity
Specifications

Specifications

TAGLN
TAGLN/247
Mouse monoclonal antibody raised against full length recombinant human TAGLN.
In 10 mM PBS.
Liquid
TAGLN
Recombinant protein corresponding to full length human TAGLN.
Protein A/G purification
RUO
Primary
6876
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q01995
DKFZp686P11128|SM22|SMCC|TAGLN1|WS3-10
Mouse
IgG1, kappa
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody recognizes a 22 kDa protein, identified as TAGLN, also designated SM22-alpha. It may cross-react with SM22-beta.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)