Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAGLN Rabbit anti-Human,Mouse,Rat,Bovine, KHL, Clone: 1D10, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen TAGLN
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB121125MI
View Documents
MilliporeSigma
ZRB121125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB12114XMI
View Documents
MilliporeSigma
ZRB12114X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

TAGLN Monoclonal antibody specifically detects TAGLN in Human,Mouse,Rat,Bovine samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

TAGLN
KHL
Rabbit
Transgelin;22 kDa actin-binding protein;Protein WS3-10;Smooth muscle protein 22-alpha;SM22-alpha
KLH-conjugated linear peptide corresponding to 12 amino acids from the C-terminal region of human Transgelin.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Purified
Human, Mouse, Rat, Bovine
TAGLN, SM22, WS3-10
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit