Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TAGLN3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549007
Description
Predicted to react with bovine, monkey and rat based on sequence homology.
Specifications
TAGLN3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9UI15, Q9R1Q8 | |
TAGLN3 | |
KLH conjugated synthetic peptide between 2-29 amino acids from the N-terminal region of human TAGLN3. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TAGLN3 | |
Transgelin-3, Neuronal protein 22, NP22, Neuronal protein NP25, TAGLN3, NP25 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29114, 56370 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction