Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ISM2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550844

Catalog No. PA550844


Add to cart

Description

Description

The antibody detects endogenous levels of total TAIL1 protein.

The protein encoded by this gene contains a type 1 thrombospondin domain, which is present in thrombospondin, a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Two alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications

Specifications

ISM2
Polyclonal
Unconjugated
ISM2
Liquid
ISM2
Synthetic peptide corresponding to residues near the C terminal of human isthmin 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
3 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q6H9L7
TAIL1, THSD3
Rabbit
IgG
100 μL
-20°C
Primary
145501
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit