Learn More
BioTek™ Take3™ and Take3 Trio Micro-Volume Microplates PROMO
Take3 and Take3 Trio Micro-Volume plates expand functionality of the BioTek microplate spectrophotometers, multi-mode and Hybrid readers, while reducing sample volumes, costs and time required for routine quantification
Manufacturer:  BioTek™ 11120571PM
Catalog No. 11-120-571PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Measure up to 16 (Take3) or 48 (Take3 Trio) 2µL samples at a time without dilution or special treatment.
  • Compatible with BioTek Eon™, Epoch™, Synergy™, PowerWave™ XS2 spectrophotometers
  • Includes Gen5™ software with pre-programmed micro-volume nucleic acid and protein quantification protocols
  • Absorbance and fluorescence detection capability
  • Easy maintenance: simply wipe-off sample locations
  • Robust design
  • Take3 Trio has two locations for BioCell™ measurements
  • Read microspots, BioCells or standard cuvets
Take3 (11-120-571) offers:
  • 16 microspots for 2µL samples; two locations for proprietary BioCell measurements (BioCell optional) and one location for standard cuvette measurements
  • Take3 Trio (11-120-613) offers:
    • 48 microspots for 2µL samples; two locations for proprietary BioCell measurements (BioCell optional)
    Nucleic acid and protein quantification, in-situ fluorometric protein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples
    Specifications
    Specifications
    Nucleic acid and protein quantification, in-situ fluorometric Pprotein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples
    BioTek Eon™, Epoch™, Synergy™, PowerWave™ XS2 readers
    2 μL
    16