Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAL-1 Mouse anti-Human, Clone: 2TL242, eBioscience™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen 14910182

Catalog No. 50-112-2758


Add to cart

Description

Description

Description: The monoclonal antibody 2TL242 recognizes human TAL-1. The TAL-1 protein is encoded by the tal-1 (T-acute leukemia or stem cell leukemia (SCL)) gene and functions as a 42 kDa basic helix loop helix transcription factor involved in mediating cell growth and differentation. Rearrangements of the tal-1 gene are common in human T-cell acute leukemias (T-ALLs) and result in aberrant transcription in leukemic cells. TAL-1 expression in leukemic cells is localized to the nucleus although both nuclear and cytoplasmic expression has been shown in normal cells such as erythroid precursors, megakaryocytes, endothelial cells, and macrophages scattered throughout tissues such as the tonsil, cerebellum, kidney, lung and spleen. TAL-1 is known to regulate both embryonic and adult hematopoiesis and more recently was found to regulate proliferation and differentiation of common myeloid precursors to monocytes and ultimately macrophages within tissue. Applications Reported: This 2TL242 antibody has been reported for use in western blotting, microscopy, immunohistochemical staining, and immunocytochemistry. Applications Tested: This 2TL242 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using either low or high pH antigen retrieval at less than or equal to 20 µg/mL. This 2TL242 antibody has also been tested by western blot of reduced Jurkat cell lysate at less than or equal to 5 µg/mL.

It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered. TAL-1 is a basic helix-loop-helix transcription. It regulates differentiation and survival during hemopoiesis. TAL-1 is also implicated in the genesis of hemopoietic maligcies and it may play an important role in hemopoietic differentiation. It serves as a positive regulator of eryhtroid differentiation. Mutations are associated with T-cell leukemia and melanoma. TAL-1 is know to bind to the LIM domain containing protein Rhombotin.
Specifications

Specifications

TAL-1
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P17542
SCL, TCL5, bHLHa17, tal-1
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2TL242
Unconjugated
TAL1
Liquid
TAL1
IgG1, kappa
100 μg
4° C
Primary
6886
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.