Learn More
Troemner™ Talboys™ Advanced Digital 1000MP Microplate Shaker
Designed to be used in a variety of environmental conditions. The Troemner™ Talboys™ Advanced Digital 1000MP Microplate Shaker is ideal for immunoassays and general mircoplate shaking.
$1201.00 - $1209.00
Specifications
Amperage 5
Wattage 25
View More Specs
Products 2
Catalog Number Mfr. No. Voltage Price Quantity & Availability  
Catalog Number Mfr. No. Voltage Price Quantity & Availability  
02-217-993
View Documents
Troemner™
30531481
Each for $1,209.00
Add to cart
02-217-994
View Documents
Troemner™
30531194
Each for $1,201.00
Add to cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Description
  • Holds up to 4 microplates or 2 micro-tube racks
  • Accepts deep well plates
  • Timer with audible alarm
  • 5 year industry leading warranty
Specifications
Specifications
5
25