Learn More
Troemner™ Talboys™ Conical Bottom Centrifuge Tube Block
Constructed from a solid anodized aluminum block. The Troemner™ Talboys™ Conical Bottom Centrifuge Tube Block gives maximum heat retention.
Manufacturer:  Troemner™ 949131
 View more versions of this product
Catalog No. 02-218-038
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Close contact of tubes to block walls allow for maximum heat retention
  • Includes a thermometer well for measuring block temperature
Specifications
Specifications
Anodized aluminum
17.1mm
12
3.75 x 3 x 2 in. (9.5 x 7.6 x 5.1cm)
44.5mm
12
15ml