Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Troemner™ Talboys™ Mini Dry Block Heaters Products for Science Education

Accepts one interchangeable mini block for 0.2, 0.5, 1.5, 2, 15, or 50mL tubes.

Manufacturer:  Troemner™ 949TAMNLUS

 View more versions of this product

Catalog No. S14164

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Ergonomic and compact design
  • Clear polycarbonate lid prevents evaporation and ensures uniform temperature
  • Features close tube and block contact
  • Includes block lifting tool
  • Available with/without heated lid
  • Dimensions: 16 x 12.7 x 12.3cm (6.3 x 5 x 4.85 in.)

949TAMNLUS

  • Without second heater in the lid
  • Heated Lid Model (949TAMHLUS)
    • Includes second heater in lid to help prevent condensation

    Applications:

    LAMP assays, immunoassays, enzyme reactions and denaturations

    Warranty and Services

    5 year industry leading limited warranty on parts and labor

    Specifications

    Specifications

    Dry Bath/Block
    Education
    16 x 12.7 x 12.3 cm (6.3 x 5 x 4.85 in.)
    LAMP assays, immunoassays, enzyme reactions and denaturations
    2 min.
    ±0.2°C
    5 Years
    ±0.5°C
    0.645 A
    Ambient +5°C to 100°C
    50/60 Hz
    Without second heater in lid
    120 V
    80 w
    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit