Learn More
Troemner™ Talboys™ Professional Digital 1000-3 Incubating Orbital Shaker
Designed to heat and shake a variety of samples. The Troemner™ Talboys™ Professional Digital 1000-3 Incubating Orbital Shaker has a five year warranty on parts and labor.
Manufacturer:  Troemner™ 980184
 View more versions of this product
Catalog No. 02-217-995
  Check Availability
Add to cart
Description
Description
  • Timer with audible alarm
  • LED Displays for temperature, speed and time
  • Calibration mode for temperature
  • CSA Listed certification
  • Includes power cord