Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Troemner™ Talboys™ Standard Analog 1000 Orbital Shaker
Designed for educational labs or basic shaking applications. The Troemner™ Talboys™ Standard Analog 1000 Orbital Shaker includes a non-skid rubber mat.
Supplier: Troemner™ 30531478
Description
- Variable speed microprocessor control
- Low profile design
- 15 mm orbit
- Ideal for blotting techniques, staining/destaining and general shaking procedures
- 5 year warranty
- Made in the USA
Specifications
15 mm | |
40 to 300 rpm | |
120 V | |
25 W | |
CSA Listed | |
50/60 Hz | |
4 in. | |
17 in. | |
8.75 in. | |
Analog | |
Maximum 80%, Non-condensing |
3.6 kg | |
120 V, 5 A, 25 W | |
0°C to 40°C | |
5 A | |
32°F to 104°F | |
17 x 11 x 4 in. (43.2 x 27.9 x 10.2 cm) | |
10.2 cm | |
43.2 cm | |
22.2 cm | |
Orbital Shaker |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title