Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talc, FCC, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
14807-96-6
H2Mg3O12Si4
379.26
KNXVOGGZOFOROK-UHFFFAOYSA-N
trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O
14807-96-6
H2Mg3O12Si4
379.26
KNXVOGGZOFOROK-UHFFFAOYSA-N
trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O

T1002, 14807-96-6, 3MgO•4SiO2•H2O

Spectrum™ Talc, FCC is approved as a carrier for food colorings and as a separating agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Quantity 12 kg
Ignition Residue 0.06
Loss on Drying 0.005
CAS Max % 1
Grade FCC