Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talc, Powder, Multi-Compendial, J.T.Baker™

Click to view available options
Quantity:
500g
2kg
Packaging:
Poly Bottle

Chemical Identifiers

14807-96-6
H2Mg3O12Si4
379.259
FPAFDBFIGPHWGO-UHFFFAOYSA-N
16211421
SMILES O.O=[Mg].O=[Mg].O=[Mg].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O
IUPAC Name dioxosilane;oxomagnesium;hydrate
Packaging Poly Bottle
Physical Form Powder
Quantity 2kg
Chemical Name or Material Talc
Grade Multi-Compendial