Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talc, Powder, USP, Spectrum™

Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
14807-96-6
H2Mg3O12Si4
379.26
KNXVOGGZOFOROK-UHFFFAOYSA-N
trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O
14807-96-6
H2Mg3O12Si4
379.26
KNXVOGGZOFOROK-UHFFFAOYSA-N
trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O

T1005, 14807-96-6, 3MgO•4SiO2•H2O

Spectrum™ Talc, Powder, USP is used as a carrier for pharmaceuticals with a disinfectant, astringent, anti-itching or cooling effect. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Ignition Residue 0.07
Alkalinity 0.4 mL
Acidity 0.3 mL
Quantity 45 kg
CAS Max % 1
Grade USP