Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551129

Catalog No. PA551129


Add to cart

Description

Description

Talin Polyclonal Antibody detects endogenous levels of Talin protein.

Probably involved in connections of major cytoskeletal structures to the plasma membrane. High molecular weight cytoskeletal protein concentrated at regions of cell-substratum contact and, in lymphocytes, at cell-cell contacts. {ECO:0000250}.
Specifications

Specifications

Talin 1
Polyclonal
Unconjugated
TLN1
Liquid
TLN1
Synthesized peptide derived from the C-terminal region of human Talin-1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9Y490, P26039
KIAA1027, TLN
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
21894, 313494, 7094
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit