Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talin1 Antibody (93E12), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250319FR
Description
Talin1 Monoclonal specifically detects Talin1 in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Talin1 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TLN1 | |
Recombinant protein fragment | |
0.1 mL | |
Cytoskeleton Markers | |
7094 | |
Mouse | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
93000000000000 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ILWEQ, KIAA1027, talin 1, talin-1, TLN | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Does not cross react with Talin2. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction