Learn More
Talin1 Mouse anti-Chicken, Janelia Fluor 646, Clone: TA205, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP121642JF646
Catalog No. NBP12642646
Add to cart
Description
Description
Talin1 Monoclonal antibody specifically detects Talin1 in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
Talin1
TA205
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Purified
TLN1
Human platelet talin.
Protein G purified
Cytoskeleton Markers
Primary
7094
Chicken, Human, Rabbit
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
ILWEQ, KIAA1027, talin 1, talin-1, TLN
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Immunoblots the 225 kDa talin band on SDS gels of platelets and other cells. It may be used to stain adhesion plaques in fixed and permeabilized fibroblasts. TA205 recognizes an epitope within residues 139-433.