Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ TallPrep™ Metal Bead Lysing Matrix Tubes Available on GSA/VA Contract for Federal Government customers only.

For nucleic acid isolation and purification

$314.00 - $881.50

Specifications

Product Type Lysis Matrix Tube
For Use With (Application) Nucleic acid isolation and purification
For Use With (Equipment) FastPrep-24, FastPrep 120, FastPrep-96
Matrix Stainless-steel
Sample Type Skeletal muscle, Lung, Heart, Bone, Seeds, Bacteria (Gram +/-), Yeast, Fungi;, Spores, Plants, Fixed samples, Sludge;, Wastewater, Feces, Food, Roots, Soil, Animal tissue
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-694-5100 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MP Biomedicals™
116945100
Each for $881.50
Add to cart
 
MP116945025 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MP Biomedicals™
116945025
25 x 4.5mL
Each for $314.00
Add to cart
 
MP116945050 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MP Biomedicals™
116945050
50 x 4.5mL
Each for $584.50
Add to cart
 
Description

Description

  • Each impact-resistant 4.5 mL tube contains twelve 3.175mm (1/8 in.) diameter stainless steel beads
  • Use to break hard/brittle samples
  • Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing

Certifications

DNase and RNase free

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Lysis Matrix Tube
FastPrep-24, FastPrep 120, FastPrep-96
Skeletal muscle, Lung, Heart, Bone, Seeds, Bacteria (Gram +/-), Yeast, Fungi;, Spores, Plants, Fixed samples, Sludge;, Wastewater, Feces, Food, Roots, Soil, Animal tissue
Nucleic acid isolation and purification
Stainless-steel
Room temperature
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit