Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Talniflumate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
66898-62-2
C21H13F3N2O4
414.34
ANMLJLFWUCQGKZ-UHFFFAOYSA-N
talniflumate,somalgen,lomucin,talniflumato,talniflumate usan:inn,talniflumatum inn-latin,talniflumato inn-spanish,phthalidyl 2-3-trifluoromethylanilino nicotinate,3-phthalidyl ester of 2-3-trifluoromethyl phenyl amino-3-pyridinecarboxylic acid,phthalidyl 2-alpha,alpha,alpha-trifluoro-m-toluidino nicotinate
48229
(3-oxo-1H-2-benzofuran-1-yl) 2-[3-(trifluoromethyl)anilino]pyridine-3-carboxylate
C1=CC=C2C(=C1)C(OC2=O)OC(=O)C3=C(N=CC=C3)NC4=CC=CC(=C4)C(F)(F)F
This item is not returnable. View return policy
66898-62-2
C21H13F3N2O4
414.34
ANMLJLFWUCQGKZ-UHFFFAOYSA-N
talniflumate,somalgen,lomucin,talniflumato,talniflumate usan:inn,talniflumatum inn-latin,talniflumato inn-spanish,phthalidyl 2-3-trifluoromethylanilino nicotinate,3-phthalidyl ester of 2-3-trifluoromethyl phenyl amino-3-pyridinecarboxylic acid,phthalidyl 2-alpha,alpha,alpha-trifluoro-m-toluidino nicotinate
48229
(3-oxo-1H-2-benzofuran-1-yl) 2-[3-(trifluoromethyl)anilino]pyridine-3-carboxylate
C1=CC=C2C(=C1)C(OC2=O)OC(=O)C3=C(N=CC=C3)NC4=CC=CC(=C4)C(F)(F)F

CaCC blocker and Cl-/HCO3- exchange inhibitor

Calcium-activated chloride channel (CaCC) (hCLCA1/mCLCA3) blocker; reduces mucin synthesis and release in cell culture and animal models. Possesses anti-inflammatory actions via inhibition of cyclooxygenases and inhibits Cl -HCO 3 - exchanger activity. Increases survival in a cystic fibrosis mouse model of distal intestinal obstructive syndrome.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 414.34
Percent Purity >99%
Chemical Name or Material Talniflumate

Recommended Storage : Store at +4°C