Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Talniflumate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Calcium-activated chloride channel (CaCC) (hCLCA1/mCLCA3) blocker; reduces mucin synthesis and release in cell culture and animal models. Possesses anti-inflammatory actions via inhibition of cyclooxygenases and inhibits Cl-/HCO3- exchanger activity. Increases survival in a cystic fibrosis mouse model of distal intestinal obstructive syndrome.
Specifications
Quantity | 50 mg |
Formula Weight | 414.34 |
Percent Purity | >99% |
Chemical Name or Material | Talniflumate |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction