Learn More
Talopram hydrochloride, Tocris
Potent, selective inhibitor of noradrenalin transporters
Manufacturer:  R&D Systems 3145/10
Quantity 10mg
 View more versions of this product
Catalog No. 31-451-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
331.88
Talopram hydrochloride
C20H25NO·HCl
1,3-Dihydro-N,3,3-trimethyl-1-phenyl-1-isobenzofuranpropanamine hydrochloride
Specifications
7013-41-4
C20H25NO·HCl
1,3-Dihydro-N,3,3-trimethyl-1-phenyl-1-isobenzofuranpropanamine hydrochloride
Talopram hydrochloride
>99%
331.88
10mg
Safety and Handling
Recommended Storage : Desiccate at Room Temperature