Learn More
Talsupram hydrochloride, Tocris™
Selective inhibitor of noradrenalin transporters
Manufacturer:  R&D Systems 4352/50
Quantity 50mg
 View more versions of this product
Catalog No. 43-525-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
347.95
Talsupram hydrochloride
C20H25NS·HCl
1,3-Dihydro-N,3,3-trimethyl-1-phenylbenzo[c]thiophene-1-propanamine hydrochloride
Specifications
25487-28-9
C20H25NS·HCl
1,3-Dihydro-N,3,3-trimethyl-1-phenylbenzo[c]thiophene-1-propanamine hydrochloride
Talsupram hydrochloride
>98%
347.95
50mg
Safety and Handling
Recommended Storage : Desiccate at Room Temperature